Human recombinant Dermatopontin (from cells)
Fournisseur: ProSci
PRSI91-801EA 525 EUR InStock
PRSI91-801
Human recombinant Dermatopontin (from cells)
Protéines et Peptides
Dermatopontin, also known as Tyrosine-rich acidic matrix protein, TRAMP and DPT, is a secreted protein which belongs to the dermatopontin family. DPT is expressed in various tissues, such as fibroblasts, heart, skeletal muscle, brain and pancreas. It seems to mediate adhesion by cell surface integrin binding. DPT may serve as a communication link between the dermal fibroblast cell surface and its extracellular matrix environment. DPT can enhance TGFB1 activity through interaction with decorin. In addition, DPT accelerates collagen fibril formation, stabilizes collagen fibrils against low-temperature dissociation and inhibits cell proliferation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité