Human recombinant IL-18RAcP (from cells)
Fournisseur: ProSci
PRSI91-804EA 306 EUR InStock
PRSI91-804
Human recombinant IL-18RAcP (from cells)
Protéines et Peptides
IL-18RAP is a single-pass type I membrane protein and contains two Ig-like C2-type domains and one TIR domain, IL18RAP can be induced by IFN-alpha and IL12 in nature killer cells and T-cells. The coexpression of IL18R1 and IL18RAP is required for the activation of NF-kappa B and JNK in response to IL-18.Defects in IL18RAP are associated with Coeliac disease.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité