Human recombinant TGF beta 2 (from cells)
Fournisseur: ProSci
PRSI91-819EA 678 EUR InStock
PRSI91-819
Human recombinant TGF beta 2 (from cells)
Protéines et Peptides
TGFBR2 is a single-pass type I membrane protein and contains one protein kinase domain. TGFBR2 exsits as a heterodimeric complex with another receptor protein and binds TGF-beta. Signals triggered through the TGF-beta receptor complex prompt various responses by the cell. One such response is to inhibit cell growth and division. Based on this action, the TGF-beta receptor type 2 is sometimes called a tumor suppressor. Defects in TGFBR2 have been associated with Marfan syndrome, Loeys-Deitz aortic aneurysm syndrome, Osler-Weber-Rendu syndrome and the development of various types of tumors.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité