Human recombinant ICOS (from cells)
Fournisseur: ProSci
PRSI91-826EA 525 EUR InStock
PRSI91-826
Human recombinant ICOS (from cells)
Protéines et Peptides
Inducible T-cell costimulator, also known as activation-inducible lymphocyte immunomediatory molecule, CD278, AILIM, CVID1 and ICOS, belongs to the CD28 and CTLA4 cell surface receptor family.. ICOS contains one Ig-like V-type domain and exsits as a homodimer with disulfide-linked. ICOS is highly expressed on tonsillar T-cellsand can be induced by PMA and ionomycin, ICOS plays an important role in cell-cell signaling, immune responses, and regulation of cell proliferation. Defects in ICOS are the cause of immunodeficiency common variable type 1, which is a primary immunodeficiency characterized by antibody deficiency, hypogammaglobulinemia, recurrent bacterial infections and an inability to mount an antibody response to antige
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité