Human recombinant IL-10 Receptor beta (from Cells)
Fournisseur: ProSci
PRSI91-829EA 365 EUR InStock
PRSI91-829
Human recombinant IL-10 Receptor beta (from Cells)
Protéines et Peptides
Interleukin-10 receptor subunit beta(IL10RB), also known as Cytokine receptor class-II member 4,Cytokine receptor family 2 member 4,Interleukin-10 receptor subunit 2, belongs to the type II cytokine receptor family. IL10RB is a single- pass type I membrane protein and contains two fibronectin type-III domains. It is an accessory chain which is essential for the active interleukin 10 receptor complex. Coexpression of IL10RB and IL10RA proteins has been shown to be required for IL10-induced signal transduction. Defects in IL10RB are the cause of inflammatory bowel disease type 25 (IBD25) which is a chronic, relapsing inflammation of the gastrointestinal tract with a complex etiology.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité