Human recombinant PSG1 (from cells)
Fournisseur: ProSci
PRSI91-848EA 761 EUR InStock
PRSI91-848
Human recombinant PSG1 (from cells)
Protéines et Peptides
Pregnancy-specific beta-1-glycoprotein 1(PSBG-1 for short), also named CD66 antigen-like family member F, Fetal liver non-specific cross-reactive antigen 1/2, PSG95, Pregnancy-specific beta-1 glycoprotein C/D, is a secreted protein which belongs to the immunoglobulin superfamily, CEA family. It contains 3 Ig-like C2-type (immunoglobulin-like) domains and 1 Ig-like V-type (immunoglobulin-like) domain. The human placenta is a multihormonal endocrine organ that produces hormones, enzymes, and other molecules that support fetal survival and development.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité