Human recombinant Butyrophilin 3A3 (from Cells)
Fournisseur: ProSci
PRSI91-851EA 525 EUR InStock
PRSI91-851
Human recombinant Butyrophilin 3A3 (from Cells)
Protéines et Peptides
Human BTN3A3, also known as butyrophilin subfamily 3 member A3 and BTF3, is a Single-pass type I membrane protein which belongs to the immunoglobulin superfamily and BTN/MOG family. The butyrophilin (BTN) genes are a group of major histocompatibility complex (MHC)-associated genes that encode type I membrane proteins with 2 extracellular immunoglobulin domains and an intracellular B30.2 (PRYSPRY) domain. It can be detected in peripheral blood mononuclear cells, T-cells l, spleen and lymphocytes. BTN3A3 plays a role in T-cell responses in the adaptive immune response.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité