Human recombinant Complement Factor H-related 5 (from cells)
Fournisseur: ProSci
PRSI91-852EA 525 EUR InStock
PRSI91-852
Human recombinant Complement Factor H-related 5 (from cells)
Protéines et Peptides
Complement factor H-related protein 5(FHR-5 for short), is a secreted protein which contains 9 Sushi (CCP/SCR) domains. It is expressed by the liver and secreted in plasma. The pattern of the deposits is similar to other complement components, suggesting that FHR-5 may play a role in complement activation and regulation. Defects in CFHR5 have been found in patients with atypical hemolytic uremic syndrome and may contribute to the disease. In contrast to typical hemolytic uremic syndrome, atypical forms have a poorer prognosis, with higher death rates and frequent progression to end-stage renal disease.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité