Human recombinant IFN alpha 1 (from cells)
Fournisseur: ProSci
PRSI91-853EA 761 EUR InStock
PRSI91-853
Human recombinant IFN alpha 1 (from cells)
Protéines et Peptides
Interferon alpha-1/13(IFN-alpha-1/13 for short), also known as Interferon alpha-D, is a secreted protein which belongs to the alpha/beta interferon family. It is produced by macrophages. IFN-alpha have antiviral activities. Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase. IFN-alpha exerts a variety of other biological effects, including antitumor and immunomodulatory activities and are increasingly used clinically to treat a range of malignancies, myelodysplasias and autoimmune diseases.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité