Human recombinant Apolipoprotein A-II (from cells)
Fournisseur: ProSci
PRSI91-854EA 761 EUR InStock
PRSI91-854
Human recombinant Apolipoprotein A-II (from cells)
Protéines et Peptides
Apolipoprotein A-II(Apo-AII for short), also known as Apolipoprotein A2, is a secreted protein which belongs to the apolipoprotein A2 family. It exists as a disulfide-linked homodimer; and also can form a disulfide-linked heterodimer with APOD. APOA2 is the 2nd most abundant protein of the high density lipoprotein particles. This protein may stabilize HDL (high density lipoprotein) structure by its association with lipids, and affect the HDL metabolism. Defects in APOA2 gene might cause apolipoprotein A-II deficiency or hypercholesterolemia.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité