Human Recombinant GMCSF (from Cells)
Fournisseur: ProSci
PRSI91-857EA 553 EUR InStock
PRSI91-857
Human Recombinant GMCSF (from Cells)
Protéines et Peptides
Granulocyte-macrophage colony-stimulating factor is also known as Colony-stimulating factor,CSF, Molgramostin and Sargramostim. In humans, it is encoded by the CSF2 gene. It belongs to the GM-CSF family. Granulocyte-macrophage colony-stimulating factor is a cytokine that stimulates the growth and differentiation of hematopoietic precursor cells from various lineages, including granulocytes, macrophages, eosinophils and erythrocytes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité