Human recombinant IFN omega 1 (from cells)
Fournisseur: ProSci
PRSI91-864EA 553 EUR InStock
PRSI91-864
Human recombinant IFN omega 1 (from cells)
Protéines et Peptides
Interferon omega-1 is also known as Interferon alpha-II-1and IFNW1. It is a Secreted protein that in humans is encoded by the IFNW1 gene. IFNW1 belongs to the alpha/beta interferon family. Type I IFNs consist of IFN alpha, beta , τ, and ω and bind to the type I IFN receptor, whereas IFN- gamma is the only type II IFN and is specific for the type II IFN receptor. IFNW1 is a recently discovered protein structurally related to IFN-alpha and –beta. It has been shown that IFN-omega 1 similar to that of other human class I IFNs; potent antiviral activity was also observed on cells of bovine and ovine but not of equine or murine origin.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité