Human recombinant TWEAK receptor (from cells)
Fournisseur: ProSci
PRSI91-900EA 525 EUR InStock
PRSI91-900
Human recombinant TWEAK receptor (from cells)
Protéines et Peptides
Tumor necrosis factor receptor superfamily member 12A(TNFRSF12A) is also known as Fibroblast growth factor-inducible immediate-early response protein 14, FN14, CD266 antigen and tweak-receptor. TNFRSF12A is a single-pass type I membrane protein, including a 27 aa signal peptide, a 53 aa extracellular domain, a 21 aa transmembrane domain and a 28 aa cytoplasmic domain. TNFRSF12A is highly expressed in heart, placenta and kidney. TNFRSF12A can be induced by FGF1 and phorbol ester. TNFRSF12A binds to TWEAK/TNFSF12A to initiate a signal transduction cascade, causing different cellular responses such as cell death, cell proliferation, and angiogenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité