Mouse recombinant cathepsin E
Fournisseur: ProSci
PRSI91-901EA 761 EUR InStock
PRSI91-901
Mouse recombinant cathepsin E
Protéines et Peptides
Cathepsin E is encoded by the ctse gene, exists in the homodimer forms, belongs to the peptidase A1 family. Cathepsin E high expressed in the stomach, clara cells and alveolar macrophages of lung, brain microglia, spleen and activated B-lymphocytes. Cathepsin E may involve in the processing of antigenic peptides during MHC class II-mediated antigen presentation, play a role in activation-induced lymphocyte depletion in the thymus, and in neuronal degeneration and glial cell activation in the brain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité