Human recombinant Ephrin A1 (from cells)
Fournisseur: ProSci
PRSI91-935EA 525 EUR InStock
PRSI91-935
Human recombinant Ephrin A1 (from cells)
Protéines et Peptides
Ephrin-A1 is a member of the A-type ephrin family of cell surface proteins that function as ligands for the A-type Eph receptor tyrosine kinase family. Ephrin-A1 can be induced by TNF and IL1B. Its expression levels can be down-regulated in primary glioma tissues compared to the normal tissues. The soluble monomeric form is expressed in the glioblastoma multiforme (GBM) and breast cancer cells. Soluble Ephrin-A1 is necessary for the transformation of HeLa and SK-BR3 cells and participates in the relocalization of EPHA2 away from sites of cell-cell contact during transformation. Ephrin-A1 plays an important role in angiogenesis and tumor neovascularization.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité