Mouse recombinant GITR
Fournisseur: ProSci
PRSI91-947EA 525 EUR InStock
PRSI91-947
Mouse recombinant GITR
Protéines et Peptides
Tumor necrosis factor receptor superfamily member 18(Gitr) contains 3 TNFR-Cys repeats and it have four isforms.IsformA、isformB and isformC is single-pass type I membrane protein and isformD is a secreted protein. The protein is the receptor for TNFSF18.It seems to be involved in interactions between activated T-lymphocytes and endothelial cells and in the regulation of T-cell receptor-mediated cell death. It mediated NF-kappa-B activation via the TRAF2/NIK pathway.It binds to TRAF1, TRAF2, and TRAF3, but not TRAF5 and TRAF6 and binds through its C-terminus to SIVA1/SIVA.It preferentially expressed in activated T lymphocytes and up-regulated in peripherical mononuclear cells after antigen stimulation/lymphocyte activation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité