Human recombinant retbindin (from cells)
Fournisseur: ProSci
PRSI91-964EA 761 EUR InStock
PRSI91-964
Human recombinant retbindin (from cells)
Protéines et Peptides
Human Retbindin is a 229 amino acid secreted protein that belongs to the folate receptor family. The gene that encodes retbindin exists as two alternatively spliced isoforms. Retbindin is first expressed in retina. It may play a role in binding retinoids and other carotenoids as it shares homology with;riboflavin binding proteins. RTBDN gene was first identified in a study of human eye tissues.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité