Human recombinant SUMO3 (from E. coli)
Fournisseur: ProSci
PRSI91-972EA 306 EUR InStock
PRSI91-972
Human recombinant SUMO3 (from E. coli)
Protéines et Peptides
SUMO3 belongs to the SUMO protein family and operates like ubiquitin. Ubiquitin-like protein which can be covalently attached to target lysines either as a monomer or as a lysine-linked polyer. Nevertheless unlike ubiquitin that targets proteins for degration, SUMO3 takes part in several cellular processess, such as nuclear transport, transcription regulation, apoptosis and protein stability. SUMO3 participates in amyloid beta generation and has a key role in the oneset or progression of Alzheimer's disease.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité