Human recombinant beta-Actin (from E. coli)
Fournisseur: ProSci
PRSI91-982EA 789 EUR InStock
PRSI91-982
Human recombinant beta-Actin (from E. coli)
Protéines et Peptides
Actins are ubiquitous globular and highly conserved proteins that are involved in various types of cell motility, structure, and integrity. Three main groups of actin isoforms, alpha, beta and gamma have been identified. The alpha actins are found in muscle tissues and are a major constituent of the contractile apparatus. The beta and gamma actins co-exist in most cell types as components of the cytoskeleton, and as mediators of internal cell motility. ACTB is a major constituent of the contractile apparatus and one of the two nonmuscle cytoskeletal actins. Polymerization of globular actin (G-actin) leads to a structural filament (F-actin) in the form of a two-stranded helix. Each actin can bind to 4 others.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité