Human recombinant B-Cell Linker (from E. coli)
Fournisseur: ProSci
PRSI91-991EA 761 EUR InStock
PRSI91-991
Human recombinant B-Cell Linker (from E. coli)
Protéines et Peptides
B-Cell Linker Protein (BLNK) is a cell membrane protein which contains 1 SH2 domain. BLNK is expressed in B cells and fibroblast cell lines, playing a important role in B cell receptor signaling. BLNK as a central linker protein, downstream of the B-cell receptor (BCR), bridges the SYK kinase to a multitude of signaling pathways and regulating biological outcomes of B-cell function and development. BLNK associates with the activation of ERK/EPHB2, MAP kinase p38 and JNK, modulates AP1, NF-kappa-B and NFAT activation. BLNK involves in BCR-mediated PLCG1 and PLCG2 activation and Ca2+ mobilization and is required for trafficking of the BCR to late endosomes. BLNK deficiency results in agammaglobulinemia type 4 and much more profound block in B-cell development.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité