Human recombinant WW domain-binding 1 (from E. coli)
Fournisseur: ProSci
PRSI92-005EA 761 EUR InStock
PRSI92-005
Human recombinant WW domain-binding 1 (from E. coli)
Protéines et Peptides
WW Domain-Binding Protein 1 (WBP1) is widely expressed in many tissues, but it is lowly expressed in the lung, placenta, kidney, and liver. WBP1 contains two WW-binding motifs: WW-binding 1 and WW-binding 2 that are involved in mediating protein-protein interactions through the binding of polyproline ligands. The WW-binding domain is composed of 38 to 40 semi-conserved amino acids shared by proteins with diverse functions including structural, regulatory, and signaling proteins. In addition, WBP1 also encodes a ligand of the WW domain of the Yes kinase-associated protein. This function has not been determined.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité