Human recombinant BCL-2-associated Athanogene 1 (from E. coli)
Fournisseur: ProSci
PRSI92-006EA 761 EUR InStock
PRSI92-006
Human recombinant BCL-2-associated Athanogene 1 (from E. coli)
Protéines et Peptides
BAG Family Molecular Chaperone Regulator 2 (BAG2) is a member of the Bag family whose members compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. BAG2 contains 1 BAG domain and is a important component of the HSC 70/CHIP chaperone-dependent ubiquitin ligase complex. In mammalian cells BAG1, BAG2, and BAG3 bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité