Human Recombinant MORF4L2 (from E. coli)
Fournisseur: ProSci
PRSI92-009EA 761 EUR InStock
PRSI92-009
Human Recombinant MORF4L2 (from E. coli)
Protéines et Peptides
Mortality Factor 4-Like Protein 2 (MORF4L2) is a member of the mortality factor (MORF) family. MORF4L2 localizes in the nucleus, possessing a protein kinase C phosphorylation site and a tyrosine phosphorylation site. MORF4L2 interacts with the Rb tumor suppressor and it has histone deacetylase activity which can either repress or promote the activity of the B-Myb promoter depending on the tissue. In addition, MORF4L2 is involved in cell growth, regulation, and senescence.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité