Human recombinant ADPRH (from E. coli)
Fournisseur: ProSci
PRSI92-010EA 789 EUR InStock
PRSI92-010
Human recombinant ADPRH (from E. coli)
Protéines et Peptides
ADP-Ribosylarginine Hydrolase (ADPRH) belongs to the ADP-Ribosylglycohydrolase family. ADPRH catalyzes removal of mono-ADP-ribose from arginine residues of proteins in the ADP-Ribosylation cycle, which is a post-translation modification that includes the addition of one or more ADP-ribose moieties. These reactions are related to cell signaling and the control of many cell processes, such as DNA repair and cell apoptosis. In addition, ADPRH binds with magnesium ion and possess ADP-ribosylarginine hydrolase activity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité