Human recombinant UBAP1 (from E. coli)
Fournisseur: ProSci
PRSI92-012EA 761 EUR InStock
PRSI92-012
Human recombinant UBAP1 (from E. coli)
Protéines et Peptides
Ubiquitin-Associated Protein 1 (UBAP1) belongs to the UBA domain family. Members of this family are related to ubiquitin and the ubiquitination pathway. Because of their cytogenetic location, this UBA domain family member is being studied as a putative target for mutation in nasopharyngeal carcinomas. UBAP1 is highly expressed in the heart, brain, placenta, lung, skeletal muscle, liver, and pancreas. UBAP1 consists of two UBA domains and one UMA domain. The ubiquitin associated domain is throught to be a non-covalent ubiquitin binding domain, including a compact three helix bundle.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité