Human recombinant Bridging Integrator 2 (from E. coli)
Fournisseur: ProSci
PRSI92-013EA 761 EUR InStock
PRSI92-013
Human recombinant Bridging Integrator 2 (from E. coli)
Protéines et Peptides
Bridging Integrator 2 (BIN2) is a cytoplasmic protein. BIN2 contains one BAR domain and Interacts with BIN1. BIN2 is highly expressed in some hematopoietic tissues, including peripheral blood, thymus, colon and placenta. BIN2 is an Arabidopsis GSK3-like kinase that negatively regulates brassinosteroid (BR) signaling. Genetic studies show that BIN2 is inhibited in response to BR perception at the cell surface to relieve its inhibitory effects on downstream targets.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité