Human Recombinant NIP7 (from E. coli)
Fournisseur: ProSci
PRSI92-014EA 761 EUR InStock
PRSI92-014
Human Recombinant NIP7 (from E. coli)
Protéines et Peptides
60S Ribosome Subunit Biogenesis Protein NIP7 Homolog (NIP7) belongs to the NIP7 family. NIP7 contains one PUA domain, it is essential for the process of proper 27S pre-rRNA and 60S ribosome subunit assembly. NIP7 is a monomer form and interacts with NOL8 and SBDS, and may bind to RNA. In addition, NIP7 is one of the many trans-acting factors required for eukaryotic ribosome biogenesis, which interacts with nascent pre-ribosomal particles and dissociates as they complete maturation and are exported to the cytoplasm.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité