Human recombinant 4HPPD (from E. coli)
Fournisseur: ProSci
PRSI92-016EA 789 EUR InStock
PRSI92-016
Human recombinant 4HPPD (from E. coli)
Protéines et Peptides
4-Hydroxyphenylpyruvate Dioxygenase (4HPPD) belongs to the 4HPPD family. 4HPPD is a key enzyme in the degradation of tyrosine, which catalyzes the second reaction in the catabolism of tyrosine the conversation of 4-hydroxyphenylpyruvate to homogentisate. 4HPPD exists in homodimer forms, which uses zinc as a cofactor to catalyze the third step in the conversion of L-phenylalanine to fumarate and acetoacetic acid. When the active 4HPPD enzyme concentration is low in the human body, it results in high levels of tyrosine concentration in the blood, which can cause mild mental retardation at birth, and degradation in vision as a patient grows older.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Commander maintenant
COMMANDE
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité