Human recombinant Toll-Interacting (from E. coli)
Fournisseur: ProSci
PRSI92-023EA 761 EUR InStock
PRSI92-023
Human recombinant Toll-Interacting (from E. coli)
Protéines et Peptides
Toll-Interacting Protein (TOLLIP) is a member of the tollip family. TOLLIP localizes to the cytoplasm. It contains one C2 domain and one CUE domain. TOLLIP is an inhibitory adaptor protein for Toll-like receptors (TLR). The Toll-like receptors pathway is a part of the immune system that recognize structurally conserved molecular patterns of microbial pathogens, resulting in an inflammatory immune response. TOLLIP constitutes a complex with Tom1 to regulate endosomal transferring of ubiquitinated proteins. TOLLIP can negative regulate Toll-like receptors signaling, which may limit the production of proinflammatory mediators during the process of inflammation and infection.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité