Human Recombinant EIF4E-Binding 2 (from E. coli)
Fournisseur: ProSci
PRSI92-025EA 761 EUR InStock
PRSI92-025
Human Recombinant EIF4E-Binding 2 (from E. coli)
Protéines et Peptides
Eukaryotic Translation Initiation Factor 4E-Binding Protein 2 (EIF4EBP2) is a member of the Eukaryotic Translation Initiation Factor 4E Binding Protein Family. EIF4EBP2 regulates eIF4E activity by preventing its assembly into the eIF4F complex, mediates the regulation of protein translation by hormones, growth factors and other stimuli that signal through the MAP kinase pathway. This regulation of is associated to cell proliferation, cell differentiation and viral infection. Phosphorylated EIF4EBP2 on serine and threonine residues in response to insulin, EGF and PDGF.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité