Human recombinant Importin alpha 2 (from E. coli)
Fournisseur: ProSci
PRSI92-026EA 789 EUR InStock
PRSI92-026
Human recombinant Importin alpha 2 (from E. coli)
Protéines et Peptides
Karyopherin Subunit alpha-2 (KPNA2) belongs to the importin alpha family. KPNA2 is widely expressed in many tissues and contains an N-terminal hydrophilic region, a hydrophobic central region composed of 10 repeats, and a short hydrophilic C-terminus. KPNA2 interacts with the NLSs of DNA helicase Q1 and SV40 T antigen and may be involved in the nuclear transport of proteins. KPNA2 also may play a role in V(D)J recombination. KPNA2 functions in nuclear protein importantly as an adapter protein for nuclear receptor KPNB1.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité