Human Recombinant GADD45A (from E. coli)
Fournisseur: ProSci
PRSI92-028EA 761 EUR InStock
PRSI92-028
Human Recombinant GADD45A (from E. coli)
Protéines et Peptides
Growth Arrest and DNA Damage-Inducible Protein GADD45 alpha (GADD45A) is a member of the GADD45 family. GADD45A can be induced by UV irradiation, X-rays, growth arrest and alkylating agents, of which can be mediated by some kinases other than PKC. GADD45A can interact with MAPK14, GADD45GIP1, PCNA. In T-cells, GADD45A functions as a regulator of p38 MAPKs by inhibiting p88 phosphorylation and activity. GADD45A may affect PCNA interaction with some cell division protein kinase complexes, stimulating DNA excision repair in vitro and inhibits entry of cells into S phase.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité