Human Recombinant Desmin (from E. coli)
Fournisseur: ProSci
PRSI92-030EA 761 EUR InStock
PRSI92-030
Human Recombinant Desmin (from E. coli)
Protéines et Peptides
Desmin is a cytoplasmic protein and belongs to the intermediate filament family. interacts with DST and MTM1. Desmin is only expressed in vertebrates, however homologous proteins are found in many organisms. Desmin is the main intermediate filament in mature skeletal, cardiac and smooth-muscle cells. DES founctions as homopolymers to form a stable intracytoplasmic filamentous network connecting myofibrils to each other and to the plasma membrane. .Defects in DES are cause of the myopathy myofibrillar type 1, cardiomyopathy dilated type 1I, and neurogenic scapuloperoneal syndrome Kaeser type.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité