Human recombinant p21 (from E. coli)
Fournisseur: ProSci
PRSI92-035EA 761 EUR InStock
PRSI92-035
Human recombinant p21 (from E. coli)
Protéines et Peptides
Cyclin-Dependent Kinase Inhibitor 1 (CDKN1A) is a member of the CDI family. CDKN1A is widely expressed in all adult tissues, but low expressed in the brain tissue. CDKN1A can be induced by p53/TP53, mezerein and IFNB1, repressed by HDAC1. CDKN1A may be an important intermediate, by which p53/TP53 mediates its role as an inhibitor of cellular proliferation in response to DNA damage, CDKN1A can bind to and inhibit cyclin-dependent kinase activity, preventing phosphorylation of critical cyclin-dependent kinase substrates and blocking cell cycle progression.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité