Human recombinant alpha-A-Crystallin (from E. coli)
Fournisseur: ProSci
PRSI92-059EA 306 EUR InStock
PRSI92-059
Human recombinant alpha-A-Crystallin (from E. coli)
Protéines et Peptides
Alpha-Crystallin A Chain (CRYAA) belongs to the small heat shock protein (HSP20) family and can be induced by heat shock. The expression of CRYAA is preferentially restricted to the lens cell. CRYAA may contribute to the transparency and refractive index of the lens. CRYAA has chaperone-like activity, preventing aggregation of various proteins under a wide range of stress conditions. Two additional functions of CRYAA are an autokinase activity and participation in the intracellular architecture.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité