Human recombinant BAX (from E. coli)
Fournisseur: ProSci
PRSI92-060EA 761 EUR InStock
PRSI92-060
Human recombinant BAX (from E. coli)
Protéines et Peptides
Apoptosis Regulator BAX (BAX) belongs to the Bcl-2 family. BAX exists as a homodimer and is expressed in a wide variety of tissues. The Bax gene encodes different isoforms including Bax alpha (21 kDa) and Bax beta (24 kDa). Although both isoforms contain BH1, BH2 and BH3 domains, Bax beta has a unique carboxyl terminus and does not contain a hydrophobic transmembrane domain. BAX accelerates programmed cell death by binding to, and antagonizing the apoptosis repressor BCL2 or its adenovirus homolog E1B 19k protein. BAX also promotes activation of CASP3, and thereby apoptosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N, C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité