Human recombinant UBE2I (from E. coli)
Fournisseur: ProSci
PRSI92-086EA 365 EUR InStock
PRSI92-086
Human recombinant UBE2I (from E. coli)
Protéines et Peptides
SUMO-Conjugating Enzyme UBC9 (UBC9) belongs to the ubiquitin-conjugating enzyme family. UBC9 is homologous to ubiquitin-conjugating enzymes (E2s). However, instead of conjugating ubiquitin, UBC9 conjugates a ubiquitin homologue, Small Ubiquitin-Like Modifier 1 (SUMO-1). The conjugation of ubiquitin requires the activities of ubiquitin-activating (E1) and conjugating (E2) enzymes. It is suggested that UBC9 might play a role in DNA repair and perhaps even in aging.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité