Human recombinant PHLDA2 (from E. coli)
Fournisseur: ProSci
PRSI92-098EA 584 EUR InStock
PRSI92-098
Human recombinant PHLDA2 (from E. coli)
Protéines et Peptides
Pleckstrin Homology-Like Domain Family A Member 2 (PHLDA2) is a peripheral membrane protein that belongs to the PHLDA2 family. PHLDA2 is expressed in the placenta and adult prostate gland. In the placenta, it is present in all cells of the villous cytotrophoblast. PHLDA2 plays a role in regulating placenta growth. PHLDA2 may act via its PH domain that competes with other PH domain-containing proteins, thereby preventing their binding to membrane lipids.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité