Human recombinant SMAD4 (from E. coli)
Fournisseur: ProSci
PRSI92-105EA 553 EUR InStock
PRSI92-105
Human recombinant SMAD4 (from E. coli)
Protéines et Peptides
SMAD Family Member 4 (SMAD4) is a cytoplasmic protein that belongs to the Dwarfin/SMAD family. SMAD4 contains one MH1 (MAD homology 1) domain and one MH2 (MAD homology 2) domain. It is the component of the heterotrimeric SMAD2/SMAD3-SMAD4 complex that forms in the nucleus and is required for the TGF-mediated signaling. SMAD4 promotes binding of the SMAD2/SMAD4/FAST-1 complex to DNA and provides an activation function required for SMAD1 or SMAD2 to stimulate transcription. SMAD4 may act as a tumor suppressor. It positively regulates PDPK1 kinase activity by stimulating its dissociation from the 14-3-3 protein YWHAQ which acts as a negative regulator. Mutations or deletions in SMAD4 have been shown to result in pancreatic cancer, juvenile polyposis syndrome, and hereditary hemorrhagic telangiectasia syndrome.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité