Human Recombinant HPGDS (from E. coli)
Fournisseur: ProSci
PRSI92-110EA 789 EUR InStock
PRSI92-110
Human Recombinant HPGDS (from E. coli)
Protéines et Peptides
Hematopoietic Prostaglandin D Synthase (HPGDS) belongs to the GST superfamily and Sigma family. HPGDS contains one GST C-terminal domain and one GST N-terminal domain. HPGDS is highly expressed in adipose tissue, macrophages, and placenta, and it exists in the form of homodimer in living body. HPGDS is a cytosolic enzyme that isomerizes PGH(2). HPGDS is a bifunctional enzyme that catalyzes both the conversion of PGH2 to PGD2 and also shows low glutathione-peroxidase activity towards cumenehydroperoxide.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité