Human recombinant PDCD4 (from E. coli)
Fournisseur: ProSci
PRSI92-115EA 584 EUR InStock
PRSI92-115
Human recombinant PDCD4 (from E. coli)
Protéines et Peptides
Programmed Cell Death Protein 4 (PDCD4) is a member of the PDCD4 family. PDCD4 and EIF4A1 form a heterotrimer. One molecule of PDCD4 binds two molecules of EIF4A1. PDCD4 takes part in apoptosis via inhibiting translation initiation and cap-dependent translation.PDCD4 promotes colonic neoplastic transformation and tumor invasion. PDCD4 is an important target for microrna R-21 in breast cancer cells. Shortage of PDCD4 expression is associated with colorectal cancer. Overexpression of PDCD4 in carcinoid cells results in inhibition of cell proliferation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité