Human recombinant Synaptobrevin Homolog YKT6 (from E. coli)
Fournisseur: ProSci
PRSI92-127EA 789 EUR InStock
PRSI92-127
Human recombinant Synaptobrevin Homolog YKT6 (from E. coli)
Protéines et Peptides
Synaptobrevin Homolog YKT6 (YKT6) is an enzyme that belongs to the Synaptobrevin family. YKT6 contains a longin domain and a v-SNARE coiled-coil homology domain. YKT6 is highly conserved from yeast to human and it can functionally complement the loss of the yeast homolog in the yeast secretory pathway. It is a membrane associated, isoprenylated protein that functions at the endoplasmic reticulum-Golgi transport step. YKT6 is considered as one of the SNARE recognition molecules implicated in vesicular transport between secretory compartments.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité