Human Recombinant Myozenin-2 (from E. coli)
Fournisseur: ProSci
PRSI92-145EA 761 EUR InStock
PRSI92-145
Human Recombinant Myozenin-2 (from E. coli)
Protéines et Peptides
Myozenin 2 (MYOZ2) is a 264 amino acid protein that belongs to the myozenin family. MYOZ2 binds to Calcineurin, a phosphatase that is involved in calcium-dependent signal transduction in diverse cell types. MYOZ2 is one of the sarcomeric proteins and plays an important role in myofibrillogenesis and the modulation of calcineurin signalling. It may serve as intracellular binding proteins involved in linking Z line proteins such as alpha-actinin, gamma-filamin, TCAP/telethonin, LDB3/ZASP and plays an important role in the modulation of calcineurin signalling. Defects in MYOZ2 are the cause of familial hypertrophic cardiomyopathy type 16 (CMH16), a hereditary heart disorder.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité