Human recombinant UBE2R2 (from E. coli)
Fournisseur: ProSci
PRSI92-155EA 365 EUR InStock
PRSI92-155
Human recombinant UBE2R2 (from E. coli)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 R2 (UBE2R2) is a modification enzyme that belongs to the ubiquitin-conjugating enzyme family. UBE2R2 is involved in cell growth and transformation. It accepts ubiquitin from the E1 complex and catalyses its covalent attachment to other proteins. In vitro, UBE2R2 catalyses monoubiquitination and 'Lys-48'-linked polyubiquitination. It may be involved in degradation of katenin.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité