Mouse recombinant IL6 (from E. coli)
Fournisseur: ProSci
PRSI92-168EA 761 EUR InStock
PRSI92-168
Mouse recombinant IL6 (from E. coli)
Protéines et Peptides
Interleukin-6 (IL-6) is a pro-inflammatory cytokine that also has an important role in immunity. Mouse IL-6 appears to be directly involved in the responses that occur after infection and injury and may prove to be as important as IL-1 in regulating the acute phase response. Mouse IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages, and endothelial cells. It acts upon a variety of cells, including fibroblasts, myeloid progenitor cells, T cells, B cells and hepatocytes. IL-6 has a wide variety of biological functions: it plays an essential role in the final differentiation of B-cells into Ig-secreting cells, it induces myeloma and plasmacytoma growth, nerve cells differentiation in hepatocytes, and acute phase reactants.
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité