Mouse recombinant TNF Receptor I (from E. coli)
Fournisseur: ProSci
PRSI92-179EA 525 EUR InStock
PRSI92-179
Mouse recombinant TNF Receptor I (from E. coli)
Protéines et Peptides
tumour necrosis factor receptor superfamily member 1A (Tnfrsf1a) is a member of the tumour necrosis factor receptor superfamily. Tnfrsf1a is one of the major receptors for the tumour necrosis factor-alpha. It can activate the transcription factor NF-κB, mediate apoptosis, and function as a regulator of inflammation. Antiapoptotic protein BCL2-associated athanogene 4 (BAG4/SODD) and adaptor proteins TRADD and TRAF2 have been shown to interact with this receptor, and thus play regulatory roles in the signal transduction mediated by the receptor. Germline mutations of the extracellular domains of this receptor were found to be associated with the human genetic disorder called tumour necrosis factor associated periodic syndrome (TRAPS) or periodic fever syndrome
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité