Human recombinant putative polycomb group ASXL1 (from E. coli)
Fournisseur: ProSci
PRSI92-183EA 761 EUR InStock
PRSI92-183
Human recombinant putative polycomb group ASXL1 (from E. coli)
Protéines et Peptides
Putative Polycomb group protein ASXL1 involved in transcriptional regulation mediated by ligand-bound nuclear hormone receptors, such as retinoic acid receptors (RARs) and peroxisome proliferator-activated receptor gamma (PPARG). It acts as coactivator of RARA and RXRA through association with NCOA1. ASXL1 also acts as corepressor through recruitment of KDM1A and CBX5 to target genes in a cell-type specific manner; the function seems to involve differential recruitment of methylated histone H3 to respective promoters.
Fusion-Tag: N-GST tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité