Human recombinant Endoglin (from E. coli)
Fournisseur: ProSci
PRSI92-192EA 584 EUR InStock
PRSI92-192
Human recombinant Endoglin (from E. coli)
Protéines et Peptides
Endoglin is a single-pass type I membrane protein which restricted to endothelial cells in all tissues except bone marrow. Endoglin as major glycoprotein of vascular endothelium, it has been found on endothelial cells, activated macrophages, fibroblasts, and smooth muscle cells. Furthermore, Homodimer forms a heteromeric complex with the signalling receptors for transforming growth factor-beta: TGFBR1 and/or TGFBR2. It may have an important role in the binding of endothelial cells to integrins and/or other RGD receptors. Defects in ENG are the cause of hereditary hemorrhagic telangiectasia type 1 (HHT1), which is an autosomal dominant multisystemic vascular dysplasia, characterised by recurrent epistaxis, muco-cutaneous telangiectases, gastro-intestinal hemorrhage, and pulmonary (PAVM), cerebral (CAVM) and hepatic arteriovenous malformations.
Fusion-Tag: N-Trx, 6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité