Human recombinant Casein kinase I gamma-2 (from E. coli)
Fournisseur: ProSci
PRSI92-197EA 584 EUR InStock
PRSI92-197
Human recombinant Casein kinase I gamma-2 (from E. coli)
Protéines et Peptides
CSNK1G2 is a cytoplasmic protein which contains one protein kinase domain. It is amember of the CK1 Ser/Thr protein kinase family of the large casein kinase I (CKI) subfamily. CSNK1G2 participates in Wnt signalling, involves in brain development and vesicular trafficking and neurotransmitter releasing from small synaptic vesicles. It regulates fast synaptic transmission mediated by glutamate. SMAD3 phosphorylation promotes its ligand-dependent ubiquitination and subsequent proteasome degradation, thus inhibiting SMAD3-mediated TGF-beta responses.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité