Human recombinant LMCD1 (from E. coli)
Fournisseur: ProSci
PRSI92-210EA 789 EUR InStock
PRSI92-210
Human recombinant LMCD1 (from E. coli)
Protéines et Peptides
LMCD1 is transcriptional cofactor which contains a cysteine-rich domain in the N-terminal region and 2 LIM domains in the C-terminal region. It also has several potential phosphorylation and N-myristoylation sites and a single potential N-glycosylation site. LMCD1 is expressed in many tissues with highest abundance in skeletal muscle. LMCD1 restricts GATA6 function by inhibiting DNA-binding, resulting in repression of GATA6 transcriptional activation of downstream target genes. It plays a critical role in the development of cardiac hypertrophy via activation of calcineurin/nuclear factor of activated T-cells signalling pathway.
Fusion-Tag: N, C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité